Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp26b1
Entrez Gene ID
312495
Gene Name
cytochrome P450, family 26, subfamily b, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.4: 116261796 - 116278615 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181087.2 NP_851601.1 1 - 2 363 101
GenBank mRNA AY245532.2 - 1 - 2 363 101

View Full Product Details