Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc5a4
Entrez Gene ID
294341
Gene Name
solute carrier family 5 (glucose activated ion channel), member 4
Gene Aliases
Slc5a4a
Chromosome Location
Chr.20: 13415039 - 13458110 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106383.1 NP_001099853.1 1 - 2 223 58

View Full Product Details