Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdyl2
Entrez Gene ID
292044
Gene Name
chromodomain protein, Y-like 2
Gene Aliases
-
Chromosome Location
Chr.19: 49092855 - 49159228 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106189.1 NP_001099659.1 3 - 4 970 87

View Full Product Details