Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcl11a
Entrez Gene ID
305589
Gene Name
B-cell CLL/lymphoma 11A (zinc finger protein)
Gene Aliases
-
Chromosome Location
Chr.14: 108826715 - 108921197 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191683.1 NP_001178612.1 1 - 2 299 72
XM_006251586.2 XP_006251648.1 1 - 2 522 72
XM_006251588.2 XP_006251650.1 1 - 2 521 72
XM_006251594.2 XP_006251656.1 1 - 2 508 72
XM_008770430.1 XP_008768652.1 1 - 2 521 72
XM_008770431.1 XP_008768653.1 1 - 2 523 72
GenBank mRNA BC086607.1 - 1 - 2 408 72
BC158689.1 - 1 - 2 212 72

View Full Product Details