The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012039.1 | NP_001012039.1 | 11 - 12 | 1430 | 111 |
XM_008770456.1 | XP_008768678.1 | 11 - 12 | 1530 | 111 | |
XM_008770457.1 | XP_008768679.1 | 10 - 11 | 1450 | 111 | |
GenBank mRNA | BC079455.1 | - | 11 - 12 | 1430 | 111 |
D89730.1 | - | 9 - 10 | 1326 | 111 |