Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lcn5
Entrez Gene ID
29552
Gene Name
lipocalin 5
Gene Aliases
Erabp
Chromosome Location
Chr.3: 2876228 - 2881305 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024136.1 NP_077050.1 5 - 6 561 90
XM_008761580.1 XP_008759802.1 5 - 6 1626 90
GenBank mRNA M12790.1 - 5 - 6 513 90
X59832.1 - 5 - 6 553 90

View Full Product Details