Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
H2afy
Entrez Gene ID
29384
Gene Name
H2A histone family, member Y
Gene Aliases
-
Chromosome Location
Chr.17: 9282106 - 9344698 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017182.2 NP_058878.1 5 - 6 770 85
XM_006253573.2 XP_006253635.1 5 - 6 808 85
XM_006253574.2 XP_006253636.1 5 - 6 898 85
XM_006253577.2 XP_006253639.1 5 - 6 808 85
GenBank mRNA BC089093.1 - 5 - 6 770 85
M99066.1 - 1 - 2 102 85
U79139.1 - 5 - 6 715 85

View Full Product Details