Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr86
Entrez Gene ID
311956
Gene Name
WD repeat domain 86
Gene Aliases
RGD1566180
Chromosome Location
Chr.4: 6942415 - 6948912 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110489.1 NP_001103959.1 1 - 2 259 59

View Full Product Details