The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077660.2 | NP_001071128.1 | 5 - 6 | 230 | 70 |
XM_006251312.2 | XP_006251374.1 | 5 - 6 | 691 | 70 | |
XM_006251313.2 | XP_006251375.1 | 5 - 6 | 520 | 70 | |
XM_006251315.2 | XP_006251377.1 | 5 - 6 | 323 | 70 | |
XM_008770307.1 | XP_008768529.1 | 4 - 5 | 212 | 70 | |
GenBank mRNA | CB720935.1 | - | 3 - 4 | 108 | 70 |
CB757259.1 | - | 5 - 6 | 256 | 70 | |
CB809919.1 | - | 5 - 6 | 224 | 70 | |
FM044330.1 | - | 5 - 6 | 230 | 70 | |
FM046879.1 | - | 4 - 5 | 180 | 70 | |
FQ222248.1 | - | 5 - 6 | 222 | 70 | |
FQ223767.1 | - | 5 - 6 | 217 | 70 |