The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001030032.1 | NP_001025203.1 | 2 - 3 | 341 | 62 |
XM_006244089.2 | XP_006244151.1 | 2 - 3 | 2242 | 62 | |
GenBank mRNA | BC099762.1 | - | 2 - 3 | 341 | 62 |