The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108630.1 | NP_001102100.1 | 2 - 3 | 273 | 115 |
GenBank mRNA | BC168709.1 | - | 2 - 3 | 266 | 115 |