Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fscn1
Entrez Gene ID
683788
Gene Name
fascin actin-bundling protein 1
Gene Aliases
Fascin
Chromosome Location
Chr.12: 13655459 - 13668595 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100806.1 NP_001094276.1 - 1071 89
GenBank mRNA BC168678.1 - - 1071 89

View Full Product Details