Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
A1bg
Entrez Gene ID
140656
Gene Name
alpha-1-B glycoprotein
Gene Aliases
C44
Chromosome Location
Chr.7: 102294805 - 102298522 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022258.2 NP_071594.2 5 - 6 977 114
GenBank mRNA AF236054.1 - 1 - 2 296 114
AJ302031.1 - 5 - 6 977 114
BC089771.1 - 5 - 6 960 114

View Full Product Details