The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017466.2 | NP_001017466.2 | 3 - 4 | 595 | 141 |
GenBank mRNA | BC092576.1 | - | 3 - 4 | 595 | 141 |