The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008507.1 | NP_001008507.1 | 6 - 7 | 786 | 158 |
XM_006246472.2 | XP_006246534.1 | 7 - 8 | 994 | 158 | |
XM_006246473.2 | XP_006246535.1 | 6 - 7 | 1655 | 158 | |
XM_006246474.2 | XP_006246536.1 | 6 - 7 | 1123 | 158 | |
XM_006246475.2 | XP_006246537.1 | 5 - 6 | 806 | 158 | |
XM_008767786.1 | XP_008766008.1 | 6 - 7 | 982 | 158 | |
GenBank mRNA | BC086453.1 | - | 6 - 7 | 786 | 158 |
EF445633.1 | - | 7 - 8 | 898 | 158 |