Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lgi3
Entrez Gene ID
306013
Gene Name
leucine-rich repeat LGI family, member 3
Gene Aliases
-
Chromosome Location
Chr.15: 52220578 - 52227713 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107277.1 NP_001100747.1 5 - 6 872 91
XM_006252273.2 XP_006252335.1 4 - 5 791 91

View Full Product Details