The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109055.1 | NP_001102525.1 | 2 - 3 | 839 | 75 |
XM_006248459.2 | XP_006248521.1 | 2 - 3 | 942 | 75 | |
XM_006248460.2 | XP_006248522.1 | 2 - 3 | 743 | 75 | |
XM_006248461.2 | XP_006248523.1 | 2 - 3 | 577 | 75 |