Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtf2ird2
Entrez Gene ID
288606
Gene Name
GTF2I repeat domain containing 2
Gene Aliases
RGD1307435, RGD1566086
Chromosome Location
Chr.12: 25509059 - 25546200 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271485.1 NP_001258414.1 12 - 13 991 65
XM_008769131.1 XP_008767353.1 9 - 10 1047 65

View Full Product Details