Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rhod
Entrez Gene ID
293660
Gene Name
ras homolog family member D
Gene Aliases
-
Chromosome Location
Chr.1: 219668352 - 219682287 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106323.1 NP_001099793.1 4 - 5 558 108
GenBank mRNA FQ229623.1 - 4 - 5 507 108

View Full Product Details