Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xrcc6bp1
Entrez Gene ID
299828
Gene Name
XRCC6 binding protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 70183625 - 70198655 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134466.1 NP_001127938.1 3 - 4 377 88

View Full Product Details