The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134865.1 | NP_001128337.1 | 8 - 9 | 2205 | 67 |
XM_008765548.1 | XP_008763770.1 | 8 - 9 | 2227 | 67 |