The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138849.1 | NP_620204.1 | 6 - 7 | 1162 | 67 |
XM_006230101.2 | XP_006230163.1 | 6 - 7 | 1257 | 67 | |
XM_006230102.2 | XP_006230164.1 | 6 - 7 | 1142 | 67 | |
XM_006230104.1 | XP_006230166.1 | 6 - 7 | 1356 | 67 | |
XM_008759675.1 | XP_008757897.1 | 4 - 5 | 1622 | 67 | |
GenBank mRNA | U30831.1 | - | 6 - 7 | 1162 | 67 |