Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nt5dc3
Entrez Gene ID
691922
Gene Name
5'-nucleotidase domain containing 3
Gene Aliases
-
Chromosome Location
Chr.7: 27309964 - 27365687 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134887.1 NP_001128359.1 12 - 13 1426 56
XM_006241203.2 XP_006241265.1 12 - 13 1427 56
XM_006241204.2 XP_006241266.1 12 - 13 1428 56

View Full Product Details