Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Txlna
Entrez Gene ID
682457
Gene Name
taxilin alpha
Gene Aliases
-
Chromosome Location
Chr.5: 147830514 - 147846196 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127633.1 NP_001121105.1 9 - 10 1367 126
XM_006238972.2 XP_006239034.1 9 - 10 1374 126
GenBank mRNA BC166486.1 - 9 - 10 1367 126

View Full Product Details