The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108565.1 | NP_001102035.1 | 1 - 2 | 297 | 90 |
XM_006232004.2 | XP_006232066.2 | 1 - 2 | 354 | 90 | |
XM_008761515.1 | XP_008759737.1 | 1 - 2 | 353 | 90 | |
GenBank mRNA | BC159417.1 | - | 1 - 2 | 289 | 90 |