The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012889.1 | NP_037021.1 | - | 2112 | 98 |
GenBank mRNA | BC089812.1 | - | - | 2130 | 98 |
M84488.1 | - | - | 2112 | 98 | |
X63722.1 | - | - | 2061 | 98 |