Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Peg3
Entrez Gene ID
103691034
Gene Name
paternally expressed 3
Gene Aliases
-
Chromosome Location
Chr.1: 70210810 - 70218201 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CB711252.1 - - 95
CB743994.1 - - 301 92
CB766724.1 - - 271 92

View Full Product Details