Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btn1a1
Entrez Gene ID
306956
Gene Name
butyrophilin, subfamily 1, member A1
Gene Aliases
-
Chromosome Location
Chr.17: 43899565 - 43908986 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170337.1 NP_001163808.1 2 - 3 436 98
XM_006253970.2 XP_006254032.1 3 - 4 815 98
GenBank mRNA AY327120.1 - 2 - 3 440 98

View Full Product Details