Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tarm1
Entrez Gene ID
499065
Gene Name
T cell-interacting, activating receptor on myeloid cells 1
Gene Aliases
RGD1564272
Chromosome Location
Chr.1: 64246775 - 64256149 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008758841.1 XP_008757063.1 3 - 4 666 83

View Full Product Details