The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008825.1 | NP_001008825.1 | 8 - 9 | 1373 | 159 |
XM_006242347.2 | XP_006242409.1 | 8 - 9 | 1458 | 159 |