The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025017.1 | NP_001020188.1 | 3 - 4 | 496 | 129 |
GenBank mRNA | BC093385.1 | - | 3 - 4 | 496 | 129 |