Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam96b
Entrez Gene ID
680987
Gene Name
family with sequence similarity 96, member B
Gene Aliases
RGD1311762
Chromosome Location
Chr.19: 551708 - 553573 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144854.1 NP_001138326.1 2 - 3 268 72

View Full Product Details