Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc38a7
Entrez Gene ID
291840
Gene Name
solute carrier family 38, member 7
Gene Aliases
-
Chromosome Location
Chr.19: 9622594 - 9638221 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003705.2 NP_001003705.1 8 - 9 1365 62
XM_006255067.2 XP_006255129.1 10 - 11 1512 62
GenBank mRNA AY294328.1 - 9 - 10 1460 62
BC086369.1 - 8 - 9 1365 62

View Full Product Details