The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106025.1 | NP_001099495.1 | 6 - 7 | 1233 | 135 |
GenBank mRNA | FQ215928.1 | - | 4 - 5 | 880 | 135 |