Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam9b
Entrez Gene ID
100360496
Gene Name
family with sequence similarity 9, member B
Gene Aliases
-
Chromosome Location
Chr.X: 73496671 - 73509023 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257136.2 XP_006257198.1 4 - 5 349 88

View Full Product Details