Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam9b
Entrez Gene ID
100360496
Gene Name
family with sequence similarity 9, member B
Gene Aliases
-
Chromosome Location
Chr.X: 73496671 - 73509023 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257136.2 XP_006257198.1 6 - 7 558 89

View Full Product Details