The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270598.1 | NP_001257527.1 | 4 - 5 | 423 | 60 |
NM_001270599.1 | NP_001257528.1 | 4 - 5 | 423 | 60 | |
GenBank mRNA | FQ222567.1 | - | 4 - 5 | 191 | 60 |