The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031068.1 | NP_112330.1 | 11 - 12 | 1252 | 99 |
GenBank mRNA | AB292042.1 | - | 11 - 12 | 1176 | 99 |
AB292043.1 | - | 11 - 12 | 1165 | 99 | |
AF307852.1 | - | 2 - 3 | 169 | 99 | |
BC098014.1 | - | 11 - 12 | 1252 | 99 | |
FQ212256.1 | - | 11 - 12 | 1454 | 99 |