Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myoz3
Entrez Gene ID
689274
Gene Name
myozenin 3
Gene Aliases
-
Chromosome Location
Chr.18: 55843275 - 55861214 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001070223.5 XP_001070223.1 4 - 5 715 73

View Full Product Details