Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bace2
Entrez Gene ID
288227
Gene Name
beta-site APP-cleaving enzyme 2
Gene Aliases
-
Chromosome Location
Chr.11: 37798397 - 37882076 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002802.1 NP_001002802.1 8 - 9 1292 64
XM_006248153.2 XP_006248215.1 8 - 9 1346 64
GenBank mRNA BC166597.1 - 8 - 9 1306 64

View Full Product Details