Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nlrp4a
Entrez Gene ID
292545
Gene Name
NLR family, pyrin domain containing 4A
Gene Aliases
RGD1561918
Chromosome Location
Chr.1: 69326303 - 69443748 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106220.1 NP_001099690.1 1 - 2 313 77
XM_008758899.1 XP_008757121.1 2 - 3 402 77

View Full Product Details