The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198741.2 | NP_942036.1 | 3 - 4 | 703 | 62 |
GenBank mRNA | BC091107.1 | - | 3 - 4 | 686 | 62 |
KC222882.1 | - | 3 - 4 | 648 | 62 | |
KC222883.1 | - | 3 - 4 | 648 | 62 | |
KC222886.1 | - | 3 - 4 | 648 | 62 | |
Z49761.1 | - | 3 - 4 | 62 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006256035.2 | XP_006256097.1 | 3 - 4 | 714 | 62 |