Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tlr2
Entrez Gene ID
310553
Gene Name
toll-like receptor 2
Gene Aliases
-
Chromosome Location
Chr.2: 182840171 - 182846061 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008761102.1 XP_008759324.1 2 - 3 409 66

View Full Product Details