Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc94
Entrez Gene ID
501285
Gene Name
coiled-coil domain containing 94
Gene Aliases
-
Chromosome Location
Chr.9: 11098914 - 11108805 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109673.1 NP_001103143.1 3 - 4 303 82
XM_006244336.2 XP_006244398.1 3 - 4 367 82
GenBank mRNA BC129098.1 - 3 - 4 318 82
BC169032.1 - 3 - 4 303 82

View Full Product Details