The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127502.1 | NP_001120974.1 | 3 - 3 | 765 | 126 |
XM_006257281.2 | XP_006257343.1 | 3 - 3 | 804 | 126 | |
XM_008773438.1 | XP_008771660.1 | 3 - 3 | 785 | 126 | |
GenBank mRNA | BC166897.1 | - | 3 - 3 | 765 | 126 |
FQ215245.1 | - | 3 - 3 | 693 | 126 | |
FQ232216.1 | - | 3 - 3 | 739 | 125 |