Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec4a
Entrez Gene ID
474143
Gene Name
C-type lectin domain family 4, member A
Gene Aliases
Clecsf6, Dcir1
Chromosome Location
Chr.4: 156050040 - 156067518 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005899.2 NP_001005899.2 5 - 6 591 114
GenBank mRNA AY363176.1 - 5 - 6 637 114
AY494060.1 - 5 - 6 590 114
AY581051.1 - 5 - 6 591 114

View Full Product Details