Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RT1-M1-2
Entrez Gene ID
414786
Gene Name
RT1 class I, locus M1, gene 2
Gene Aliases
-
Chromosome Location
Chr.20: 2408026 - 2412023 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008849.1 NP_001008849.1 1 - 2 69 114
XM_006255884.2 XP_006255946.1 2 - 3 491 114

View Full Product Details