Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DTD1
Entrez Gene ID
851307
Gene Name
Gene Aliases
YDL219W
Chromosome Location
Chr.IV: 65242 - 65765 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001180279.1 NP_010062.1 1 - 2 61 93

View Full Product Details