Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPS3
Entrez Gene ID
853434
Gene Name
Gene Aliases
NEP98, YJL018W, YJL019W
Chromosome Location
Chr.X: 402897 - 404945 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181453.3 NP_012515.4 1 - 1 1949 66

View Full Product Details