Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NFT1
Entrez Gene ID
853979
Gene Name
Gene Aliases
YKR104W
Chromosome Location
Chr.XI: 656836 - 657756 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179894.1 NP_013030.1 1 - 1 852 97

View Full Product Details